missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Monoglyceride lipase Protein
A DNA sequence encoding the Mus musculus (Mouse) Monoglyceride lipase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9271-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001159721.1 | |
Monoglyceride lipase Protein | |
Research Use Only | |
Mgll | |
Recombinant Protein | |
Yeast | |
MPEASSPRRT PQNVPYQDLP HLVNADGQYL FCRYWKPSGT PKALIFVSHG AGEHCGRYDE LAHMLKGLDM LVFAHDHVGH GQSEGERMVV SDFQVFVRDV LQHVDTIQKD YPDVPIFLLG HSMGGAISIL VAAERPTYFS GMVLISPLVL ANPESASTLK VLAAKLLNFV LPNMTLGRID SSVLSRNKSE VDLYNSDPLV CRAGLKVCFG IQLLNAVARV ERAMPRLTLP FLLLQGSADR LCDSKGAYLL MESSRSQDKT LKMYEGAYHV LHRELPEVTN SVLHEVNSWV SHRIAAAGAG CPP | |
Greater than 90% as determined by SDS-PAGE. |
23945 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |