missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse MMP-3 Protein

A DNA sequence encoding the Mus musculus (Mouse) MMP-3 Protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8900-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16077081

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_034939.1
MMP-3 Protein
Research Use Only
MMP3
Recombinant Protein
E. coli
PGSPKWRKSH ITYRIVNYTP DLPRQSVDSA IEKALKVWEE VTPLTFSRIS EGEADIMISF AVGEHGDFVP FDGPGTVLAH AYAPGPGING DAHFDDDERW TEDVTGTNLF LVAAHELGHS LGLYHSAKAE ALMYPVYKSS TDLSRFHLSQ DDVDGIQSLY GTPTASPDVL VVPTKSNSLE PETSPMCSST LFFDAVSTLR GEVLFFKDRH FWRKSLRTPE PEFYLISSFW PSLPSNMDAA YEVTNRDTVF IFKGNQFWAI RGHEELAGYP KSIHTLGLPA TVKKIDAAIS NKEKRKTYFF VEDKYWRFDE KKQSMEPGFP RKIAEDFPGV DSRVDAVFEA FGFLYFFSGS SQLEFDPNAK KVTHILKSNS WFNC
Greater than 90% as determined by SDS-PAGE.
17392
500 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers