missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Meteorin-like Protein
A DNA sequence encoding the Mus musculus (Mouse) Meteorin-like protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7990-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_659046.1 | |
Meteorin-like Protein | |
Research Use Only | |
METRNL | |
Recombinant Protein | |
E. coli | |
QYSSDLCSWK GSGLTREARS KEVEQVYLRC SAGSVEWMYP TGALIVNLRP NTFSPAQNLT VCIKPFRDSS GANIYLEKTG ELRLLVRDIR GEPGQVQCFS LEQGGLFVEA TPQQDISRRT TGFQYELMSG QRGLDLHVLS APCRPCSDTE VLLAICTSDF VVRGFIEDVT HVPEQQVSVI YLRVNRLHRQ KSRVFQPAPE DSGHWLGHVT TLLQCGVRPG HGEFLFTGHV HFGEAQLGCA PRFSDFQRMY RKAEEMGINP CEINME | |
Greater than 90% as determined by SDS-PAGE. |
210029 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |