missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Mast cell protease 4 Protein

A DNA sequence encoding the Mus musculus (Mouse) Mast cell protease 4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7101-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16017380

  • 2985.00 EUR / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_034909.2
Mast cell protease 4 Protein
Research Use Only
Mcpt4
Recombinant Protein
E. coli
IIGGVESRPH SRPYMAHLEI TTERGFTATC GGFLITRQFV MTAAHCSGRE ITVTLGAHDV SKTESTQQKI KVEKQIVHPK YNFYSNLHDI MLLKLQKKAK ETPSVNVIPL PRPSDFIKPG KMCRAAGWGR TGVTEPTSDT LREVKLRIMD KEACKNYWHY DYNLQVCVGS PRKKRSAYKG DSGGPLLCAG VAHGIVSYGR GDAKPPAVFT RISSYVPWIN RVIKGE
Greater than 90% as determined by SDS-PAGE.
17227
1 mg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers