missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Mast cell protease 4 Protein
A DNA sequence encoding the Mus musculus (Mouse) Mast cell protease 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7101-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_034909.2 | |
Mast cell protease 4 Protein | |
Research Use Only | |
Mcpt4 | |
Recombinant Protein | |
E. coli | |
IIGGVESRPH SRPYMAHLEI TTERGFTATC GGFLITRQFV MTAAHCSGRE ITVTLGAHDV SKTESTQQKI KVEKQIVHPK YNFYSNLHDI MLLKLQKKAK ETPSVNVIPL PRPSDFIKPG KMCRAAGWGR TGVTEPTSDT LREVKLRIMD KEACKNYWHY DYNLQVCVGS PRKKRSAYKG DSGGPLLCAG VAHGIVSYGR GDAKPPAVFT RISSYVPWIN RVIKGE | |
Greater than 90% as determined by SDS-PAGE. |
17227 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |