Learn More
enQuireBio™ Recombinant Mouse Major urinary protein 11 Protein
A DNA sequence encoding the Mus musculus (Mouse) Major urinary protein 11, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7360-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001157998.1 | |
Major urinary protein 11 Protein | |
Research Use Only | |
Mup11 | |
Recombinant Protein | |
E. coli | |
REKINGEWHT IILASDKREK IEDNGNFRLF LEQIHVLENS LVLKFHTVRD EECSELSMVA DKTEKAGEYS VTYDGFNTFT IPKTDYDNFL MAHLINEKDG ETFQLMGLYG REPDLSSDIK ERFAQLCEEH GILRENIIDL SNANRCLQAR E | |
Greater than 81.5% as determined by SDS-PAGE. |
100039028 | |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |