missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Major urinary protein 11 Protein

A DNA sequence encoding the Mus musculus (Mouse) Major urinary protein 11, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7360-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048440

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001157998.1
Major urinary protein 11 Protein
Research Use Only
Mup11
Recombinant Protein
E. coli
REKINGEWHT IILASDKREK IEDNGNFRLF LEQIHVLENS LVLKFHTVRD EECSELSMVA DKTEKAGEYS VTYDGFNTFT IPKTDYDNFL MAHLINEKDG ETFQLMGLYG REPDLSSDIK ERFAQLCEEH GILRENIIDL SNANRCLQAR E
Greater than 81.5% as determined by SDS-PAGE.
100039028
500 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers