missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Major urinary protein 1
A DNA sequence encoding the Mus musculus (Mouse) Major urinary protein 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7038-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
Major urinary protein 1 | |
Research Use Only | |
Mup2 | |
Recombinant Protein | |
E. coli | |
EEASSTGRNF NVEKINGEWH TIILASDKRE KIEDNGNFRL FLEQIHVLEK SLVLKFHTVR DEECSELSMV ADKTEKAGEY SVTYDGFNTF TIPKTDYDNF LMAHLINEKD GETFQLMGLY GREPDLSSDI KERFAKLCEE HGILRENIID LSNANRCLQA RE | |
Greater than 90% as determined by SDS-PAGE. |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |