missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Lysyl oxidase homolog 1 Protein

A DNA sequence encoding the Mus musculus (Mouse) Lysyl oxidase homolog 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6316-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16022420

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_034859.2
Lysyl oxidase homolog 1 Protein
Research Use Only
Loxl1
Recombinant Protein
E. coli
RQAPSLPLPG RVGSDTVRGQ TRHPFGFGQV PDNWREVAVG DSTGMARART SVSQQRHGGS ASSSVSASAF ATTYRQPSPY PQQFPYPQAP FVNQYENYDP ASRTYEQGYV YYRGAGGGMG AGAAAVASAG VIYPFQPRAR YEDYGGGGGE EQPEYPAQGF YPAPERPYVP QPQPQPQPQP QPQPQPSDGL DRRYSHSLYN EGTPGFEQAY PDPSTDVSQA PAGAGGTYGG AGDPRLGWYP PYAANVPPEA YVPPRAVEPQ PPFRVLEPPY LPVRSSDAPS QGGERNGAQQ GRLSVGSVYR PNQNGRGLPD LVPDPNYVQA STYVQRAHLY SLRCAAEEKC LASTAYAPEA TDYDLRVLLR FPQRVKNQGT ADFLPNRPRH TWEWHSCHQH YHSMDEFSHY DLLDASTGKK VAEGHKASFC LEDSTCDFGN LKRYACTSHT QGLSPGCYDT YNADIDCQWI DITDVQPGNY ILKVHVNPKY IVLESDFTNN VVRCNIHYTG RYVSTTNCKI VQS
Greater than 90% as determined by SDS-PAGE.
16949
10 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers