missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse LY75 / DEC-205 / CD205 Protein
A DNA sequence encoding the Mus musculus (Mouse) LY75 / DEC-205 / CD205, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6325-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_038853.2 | |
LY75 / DEC-205 / CD205 Protein | |
Research Use Only | |
Ly75 | |
Recombinant Protein | |
E. coli | |
SESSGNDPFT IVHENTGKCI QPLSDWVVAQ DCSGTNNMLW KWVSQHRLFH LESQKCLGLD ITKATDNLRM FSCDSTVMLW WKCEHHSLYT AAQYRLALKD GYAVANTNTS DVWKKGGSEE NLCAQPYHEI YTRDGNSYGR PCEFPFLIGE TWYHDCIHDE DHSGPWCATT LSYEYDQKWG ICLL | |
Greater than 90% as determined by SDS-PAGE. |
17076 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |