missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Lithostathine-1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Lithostathine-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7183-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_033068.1 | |
Lithostathine-1 Protein | |
Research Use Only | |
Reg1 | |
Recombinant Protein | |
E. coli | |
QEAEEDLPSA RISCPEGSNA YSSYCYYFTE DRLTWADADL FCQNMNSGYL VSVLSQAEGN FVASLIKESG TTDANVWTGL HDPKRNRRWH WSSGSLFLYK SWATGSPNSS NRGYCVSLTS NTGYKKWKDD NCDAQYSFVC KFKG | |
Greater than 90% as determined by SDS-PAGE. |
19692 | |
500 μg | |
Not determined. | |
Full Length of Mature Protein | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |