missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Lingo1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Lingo1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8168-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001298005.1 | |
Lingo1 Protein | |
Research Use Only | |
Lingo1 | |
Recombinant Protein | |
Yeast | |
PPRCECSAQD RAVLCHRKRF VAVPEGIPTE TRLLDLGKNR IKTLNQDEFA SFPHLEELEL NENIVSAVEP GAFNNLFNLR TLGLRSNRLK LIPLGVFTGL SNLTKLDISE NKIVILLDYM FQDLYNLKSL EVGDNDLVYI SHRAFSGLNS LEQLTLEKCN LTSIPTEALS HLHGLIVLRL RHLNINAIRD YSFKRLYRLK VLEISHWPYL DTMTPNCLYG LNLTSLSITH CNLTAVPYLA VRHLVYLRFL NLSYNPIGTI EGSMLHELLR LQEIQLVGGQ LAVVEPYAFR GLNYLRVLNV SGNQLTTLEE SAFHSVGNLE TLILDSNPLA CDCRLLWVFR RRWRLNFNRQ QPTCATPEFV QGKEFKDFPD VLLPNYFTCR RAHIRDRKAQ QVFVDEGHTV QFVCRADGDP PPAILWLSPR KHLVSAKSNG RLTVFPDGTL EVRYAQVQDN GTYLCIAANA GGNDSMPAHL HVRSYSPDWP HQPNKTFAFI SNQPGEGEAN STRATVPFPF DIKTLIIAT | |
Greater than 80% as determined by SDS-PAGE. |
235402 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |