missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse KLK-8 / Kallikrein-8 Protein
A DNA sequence encoding the Mus musculus (Mouse) KLK-8 / Kallikrein-8, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9804-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001311327.1 | |
KLK-8 / Kallikrein-8 Protein | |
Research Use Only | |
Klk8 | |
Recombinant Protein | |
Yeast | |
ILEGRECIPH SQPWQAALFQ GERLICGGVL VGDRWVLTAA HCKKQKYSVR LGDHSLQSRD QPEQEIQVAQ SIQHPCYNNS NPEDHSHDIM LIRLQNSANL GDKVKPVQLA NLCPKVGQKC IISGWGTVTS PQENFPNTLN CAEVKIYSQN KCERAYPGKI TEGMVCAGSS NGADTCQGDS GGPLVCDGML QGITSWGSDP CGKPEKPGVY TKICRYTTWI KKTMDNRD | |
Greater than 94.7% as determined by SDS-PAGE. |
259277 | |
200 μg | |
Not determined. | |
Full. Length. of. Mature. Protein. | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |