missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Interferon epsilon Protein

A DNA sequence encoding the Mus musculus (Mouse) Interferon epsilon, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7798-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16040861

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_796322.1
Interferon epsilon Protein
Research Use Only
Ifne
Recombinant Protein
E. coli
LEPKRIPFQL WMNRESLQLL KPLPSSSVQQ CLAHRKNFLL PQQPVSPHQY QEGQVLAVVH EILQQIFTLL QTHGTMGIWE ENHIEKVLAA LHRQLEYVES LGGLNAAQKS GGSSAQNLRL QIKAYFRRIH DYLENQRYSS CAWIIVQTEI HRCMFFVFRF TTWLSRQDPD P
Greater than 90% as determined by SDS-PAGE.
230405
10 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers