missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IL7R / IL7RA Protein

A DNA sequence encoding the Mus musculus (Mouse) IL7R / IL7RA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6226-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16041740

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_032398.3
IL7R / IL7RA Protein
Research Use Only
Il7r
Recombinant Protein
E. coli
ESGNAQDGDL EDADADDHSF WCHSQLEVDG SQHLLTCAFN DSDINTANLE FQICGALLRV KCLTLNKLQD IYFIKTSEFL LIGSSNICVK LGQKNLTCKN MAINTIVKAE APSDLKVVYR KEANDFLVTF NAPHLKKKYL KKVKHDVAYR PARGESNWTH VSLFHTRTTI PQRKLRPKAM YEIKVRSIPH NDYFKGFWSE WSPSSTFETP EPKNQGGWD
Greater than 90% as determined by SDS-PAGE.
16197
500 μg
Not determined.
Extracellular Domain
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers