missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL-1 beta / IL1B Protein
A DNA sequence encoding the Mus musculus (Mouse) IL-1 beta / IL1B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8893-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_032387.1 | |
IL-1 beta / IL1B Protein | |
Research Use Only | |
IL1B | |
Recombinant Protein | |
E. coli | |
PIRQLHYRLR DEQQKSLVLS DPYELKALHL NGQNINQQVI FSMSFVQGEP SNDKIPVALG LKGKNLYLSC VMKDGTPTLQ LESVDPKQYP KKKMEKRFVF NKIEVKSKVE FESAEFPNWY ISTSQAEHKP VFLGNNSGQD IIDFTMESVS | |
Greater than 90% as determined by SDS-PAGE. |
16176 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |