missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse F-box / WD repeat-containing protein 10 Protein

A DNA sequence encoding the Mus musculus (Mouse) F-box/WD repeat-containing protein 10, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7709-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16020331

  • 2985.00 EUR / 1mg
Estimated Shipment: 26-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

F-box / WD repeat-containing protein 10 Protein
Research Use Only
Fbxw10
Recombinant Protein
E. coli
VKWQYSSDKN KVKKSKDKEE EREETSLGDE HSRSTIQGHS LKDSVSSKQE FSKSRVHLKQ TKNLSSDDME TPVGEVSHPL QKLWKVPMTP DRFLLTISAL QQAHNSEEFA YPHRPRPQVI DAWGPSIPYP RKVLSLKGKS VQHAVDQLRS SNLPTGVRQT NIPLEIQKLQ PNLKKSLHSP RVQATVPQPS LIRPKVSDSL RGDEHLTSSI DGTMRRAGPL TSMQVIKPNR MLAPRGGTAT LSPKKERPRF YTTLDPLRMN TGFMLMTVKE EKEFAEAKMK EYEASVSTKE VDPGKASKAA WIRKIKGLPI DNFMKEGKTA APELGQNVFI
Greater than 90% as determined by SDS-PAGE.
1 mg
Not determined.
Partial (see sequence information for more details).
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers