missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse F-box / WD repeat-containing protein 10 Protein
A DNA sequence encoding the Mus musculus (Mouse) F-box/WD repeat-containing protein 10, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7709-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
F-box / WD repeat-containing protein 10 Protein | |
Research Use Only | |
Fbxw10 | |
Recombinant Protein | |
E. coli | |
VKWQYSSDKN KVKKSKDKEE EREETSLGDE HSRSTIQGHS LKDSVSSKQE FSKSRVHLKQ TKNLSSDDME TPVGEVSHPL QKLWKVPMTP DRFLLTISAL QQAHNSEEFA YPHRPRPQVI DAWGPSIPYP RKVLSLKGKS VQHAVDQLRS SNLPTGVRQT NIPLEIQKLQ PNLKKSLHSP RVQATVPQPS LIRPKVSDSL RGDEHLTSSI DGTMRRAGPL TSMQVIKPNR MLAPRGGTAT LSPKKERPRF YTTLDPLRMN TGFMLMTVKE EKEFAEAKMK EYEASVSTKE VDPGKASKAA WIRKIKGLPI DNFMKEGKTA APELGQNVFI | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |