missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Elongation factor 2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Elongation factor 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9112-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_031933.1 | |
Elongation factor 2 Protein | |
Research Use Only | |
Eef2 | |
Recombinant Protein | |
Yeast | |
VNFTVDQIRA IMDKKANIRN MSVIAHVDHG KSTLTDSLVC KAGIIASARA GETRFTDTRK DEQERCITIK STAISLFYEL SENDLNFIKQ SKDGSGFLIN LIDSPGHVDF SSEVTAALRV TDGALVVVDC VSGVCVQTET VLRQAIAERI KPVLMMNKMD RALLELQLEP EELYQTFQRI VENVNVIIST YGEGESGPMG NIMIDPVLGT VGFGSGLHGW AFTLKQFAEM YVAKFAAKGE GQLSAAERAK KVEDMMKKLW GDRYFDPANG KFSKSANSPD GKKLPRTFCQ LILDPIFKVF DAIMNFRKEE TAKLIEKLDI KLDSEDKDKE GKPLLKAVMR RWLPAGDALL QMITIHLPSP VTAQKYRCEL LYEGPPDDEA AMGIKSCDPK GPLMMYISKM VPTSDKGRFY AFGRVFSGVV STGLKVRIMG PNYTPGKKED LYLKPIQRTI LMMGRYVEPI EDVPCGNIVG LVGVDQFLVK TGTITTFEHA HNMRVMKFSV SPVVRVAVEA KNPADLPKLV EGLKRLAKSD PMVQCIIEES GEHIIAGAGE LHLEICLKDL EEDHACIPIK KSDPVVSYRE TVSEESNVLC LSKSPNKHNR LYMKARPFPD GLAEDIDKGE VSARQELKAR ARYLAEKYEW DVAEARKIWC FGPDGTGPNI LTDITKGVQY LNEIKDSVVA GFQWATKEGA LCEENMRGVR FDVHDVTLHA DAIHRGGGQI IPTARRCLYA SVLTAQPRLM EPIYLVEIQC PEQVVGGIYG VLNRKRGHVF EESQVAGTPM FVVKAYLPVN ESFGFTADLR SNTGGQAFPQ CVFDHWQILP GDPFDNSSRP SQVVAETRKR KGLKEGIPAL DNFLDKL | |
Greater than 90% as determined by SDS-PAGE. |
13629 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |