missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CXCL3 / GRO gamma Protein

A DNA sequence encoding the Mus musculus (Mouse) CXCL3 / GRO gamma, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8652-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005781

  • 2985.00 EUR / 1mg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_976065.1
CXCL3 / GRO gamma Protein
Research Use Only
CXCL3
Recombinant Protein
E. coli
SELRCQCLNT LPRVDFETIQ SLTVTPPGPH CTQTEVIATL KDGQEVCLNP QGPRLQIIIK KILKSGKSS
Greater than 90% as determined by SDS-PAGE.
330122
1 mg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers