missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CXCL16 / SR-PSOX Protein

A DNA sequence encoding the Mus musculus (Mouse) CXCL16 / SR-PSOX, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8659-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005821

  • 600.00 EUR / 100µg


Estimated Shipment
06-08-2020
Add to basket

Specifications

Specifications

NP_075647.3
CXCL16 / SR-PSOX Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
66102
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Cxcl16
Recombinant Protein
E. coli
NQGSVAGSCS CDRTISSGTQ IPQGTLDHIR KYLKAFHRCP FFIRFQLQSK SVCGGSQDQW VRELVDCFER KECGTGHGKS FHHQKHLPQA STQTPEAAEG TPSDTSTPAH SQSTQHSTLP SGALSLNKEH TQPWEMTTLP SGYGLEARPE AEANEKQQDD RQQEAPGAGA ST
Videos
Documents