missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CXCL16 / SR-PSOX Protein
A DNA sequence encoding the Mus musculus (Mouse) CXCL16 / SR-PSOX, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8659-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_075647.3 | |
CXCL16 / SR-PSOX Protein | |
Research Use Only | |
Cxcl16 | |
Recombinant Protein | |
E. coli | |
NQGSVAGSCS CDRTISSGTQ IPQGTLDHIR KYLKAFHRCP FFIRFQLQSK SVCGGSQDQW VRELVDCFER KECGTGHGKS FHHQKHLPQA STQTPEAAEG TPSDTSTPAH SQSTQHSTLP SGALSLNKEH TQPWEMTTLP SGYGLEARPE AEANEKQQDD RQQEAPGAGA ST | |
Greater than 90% as determined by SDS-PAGE. |
66102 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |