missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CXCL12 / SDF-1 Protein
A DNA sequence encoding the Mus musculus (Mouse) CXCL12 / SDF-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8657-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_038683.1 | |
CXCL12 / SDF-1 Protein | |
Research Use Only | |
CXCL12 | |
Recombinant Protein | |
E. coli | |
KPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNK | |
Greater than 90% as determined by SDS-PAGE. |
20315 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |