missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CLEC4E / CLECSF9 Protein

A DNA sequence encoding the Mus musculus (Mouse) CLEC4E / CLECSF9, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8099-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16022671

  • 865.00 EUR / 100µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_064332.1
CLEC4E / CLECSF9 Protein
Research Use Only
Clec4e
Recombinant Protein
E. coli
TYRSSQISGQ NLQPHRNIKE LSCYSEASGS VKNCCPLNWK HYQSSCYFFS TTTLTWSSSL KNCSDMGAHL VVIDTQEEQE FLFRTKPKRK EFYIGLTDQV VEGQWQWVDD TPFTESLSFW DAGEPNNIVL VEDCATIRDS SNSRKNWNDI PCFYSMPWIC EMPEISPLD
Greater than 90% as determined by SDS-PAGE.
56619
100 μg
Not determined.
Extracellular Domain
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers