missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CLEC4E / CLECSF9 Protein

A DNA sequence encoding the Mus musculus (Mouse) CLEC4E / CLECSF9, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8099-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16072671

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_064332.1
CLEC4E / CLECSF9 Protein
Research Use Only
Clec4e
Recombinant Protein
E. coli
TYRSSQISGQ NLQPHRNIKE LSCYSEASGS VKNCCPLNWK HYQSSCYFFS TTTLTWSSSL KNCSDMGAHL VVIDTQEEQE FLFRTKPKRK EFYIGLTDQV VEGQWQWVDD TPFTESLSFW DAGEPNNIVL VEDCATIRDS SNSRKNWNDI PCFYSMPWIC EMPEISPLD
Greater than 90% as determined by SDS-PAGE.
56619
50 μg
Not determined.
Extracellular Domain
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers