missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CCL3 / Mip1a Protein
A DNA sequence encoding the Mus musculus (Mouse) CCL3 / Mip1a, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8644-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_035467.1 | |
CCL3 / Mip1a Protein | |
Research Use Only | |
Mip1a | |
Recombinant Protein | |
E. coli | |
APYGADTPTA CCFSYSRKIP RQFIVDYFET SSLCSQPGVI FLTKRNRQIC ADSKETWVQE YITDLELNA | |
Greater than 90% as determined by SDS-PAGE. |
20302 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |