missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CCL3 / Mip1a Protein
A DNA sequence encoding the Mus musculus (Mouse) CCL3 / Mip1a, was expressed in the hosts and tags indicated.
245.00 EUR - 2985.00 EUR
Specifications
Accession Number | NP_035467.1 |
---|---|
Gene ID (Entrez) | 20302 |
Name | CCL3 / Mip1a Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16055731
|
enQuireBio™
QP8644-EC-100UG |
100 μg |
675.00 EUR
100µg |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16075731
|
enQuireBio™
QP8644-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16065731
|
enQuireBio™
QP8644-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16085731
|
enQuireBio™
QP8644-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16095731
|
enQuireBio™
QP8644-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16005741
|
enQuireBio™
QP8644-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_035467.1 | |
CCL3 / Mip1a Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
20302 | |
Research Use Only | |
Mip1a | |
Recombinant Protein | |
E. coli | |
APYGADTPTA CCFSYSRKIP RQFIVDYFET SSLCSQPGVI FLTKRNRQIC ADSKETWVQE YITDLELNA | |
Greater than 90% as determined by SDS-PAGE. |