missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Cathepsin K Protein

A DNA sequence encoding the Mus musculus (Mouse) Cathepsin K, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5890-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15958489

  • 675.00 EUR / 100µg
Estimated Shipment: 26-09-2024
to see stock.

Description

Description

Specifications

Specifications

NP_031828.2
Cathepsin K Protein
Research Use Only
Ctsk
Recombinant Protein
E. coli
VPDSIDYRKK GYVTPVKNQG QCGSCWAFSS AGALEGQLKK KTGKLLALSP QNLVDCVTEN YGCGGGYMTT AFQYVQQNGG IDSEDAYPYV GQDESCMYNA TAKAAKCRGY REIPVGNEKA LKRAVARVGP ISVSIDASLA SFQFYSRGVY YDENCDRDNV NHAVLVVGYG TQKGSKHWII KNSWGESWGN KGYALLARNK NNACGITNMA SFPKM
Greater than 90% as determined by SDS-PAGE.
13038
100 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers