missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse C-C motif chemokine 9 Protein

A DNA sequence encoding the Mus musculus (Mouse) C-C motif chemokine 9, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8647-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005751

  • 985.00 EUR / 200µg


Estimated Shipment
13-02-2020
Add to basket

Specifications

Specifications

NP_035468.1
C-C motif chemokine 9 Protein
200 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
20308
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Ccl9
Recombinant Protein
E. coli
QITHATETKE VQSSLKAQQG LEIEMFHMGF QDSSDCCLSY NSRIQCSRFI GYFPTSGGCT RPGIIFISKR GFQVCANPSD RRVQRCIERL EQN
Videos
Documents