missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse C-C motif chemokine 25 Protein
A DNA sequence encoding the Mus musculus (Mouse) C-C motif chemokine 25, was expressed in the hosts and tags indicated.
245.00 EUR - 2645.00 EUR
Specifications
Accession Number | NP_033164.1 |
---|---|
Gene ID (Entrez) | 20300 |
Name | C-C motif chemokine 25 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16005761
|
enQuireBio™
QP8649-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16045761
|
enQuireBio™
QP8649-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16095751
|
enQuireBio™
QP8649-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16025761
|
enQuireBio™
QP8649-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16035761
|
enQuireBio™
QP8649-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16015761
|
enQuireBio™
QP8649-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_033164.1 | |
C-C motif chemokine 25 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
20300 | |
Research Use Only | |
Ccl25 | |
Recombinant Protein | |
E. coli | |
GAFEDCCLGY QHRIKWNVLR HARNYHQQEV SGSCNLRAVR FYFRQKVVCG NPEDMNVKRA MRILTARKRL VHWKSASDSQ TERKKSNHMK SKVENPNSTS VRSATLGHPR MVMMPRKTNN | |
Greater than 90% as determined by SDS-PAGE. |