missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Angiopoietin-like protein 1
A DNA sequence encoding the Mus musculus (Mouse) Angiopoietin-like protein 8, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7923-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001074409.1 | |
Angiopoietin-like protein 1 | |
Research Use Only | |
Gm6484 | |
Recombinant Protein | |
E. coli | |
VRPAPVAPLG GPEPAQYEEL TLLFHGALQL GQALNGVYRA TEARLTEAGH SLGLYDRALE FLGTEVRQGQ DATQELRTSL SEIQVEEDAL HLRAEATARS LGEVARAQQA LRDTVRRLQV QLRGAWLGQA HQEFETLKAR ADKQSHLLWA LTGHVQRQQR EMAEQQQWLR QIQQRLHTAA LPA | |
Greater than 90% as determined by SDS-PAGE. |
624219 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |