missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Alanine aminotransferase 1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Alanine aminotransferase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9872-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_877957.1 | |
Alanine aminotransferase 1 Protein | |
Research Use Only | |
GPT | |
Recombinant Protein | |
Yeast | |
ASQRNDRIQA SRNGLKGKVL TLDTMNPCVR RVEYAVRGPI VQRALELEQE LRQGVKKPFT EVIRANIGDA QAMGQRPITF FRQVLALCVY PNLLSSPDFP EDAKRRAERI LQACGGHSLG AYSISSGIQP IREDVAQYIE RRDGGIPADP NNIFLSTGAS DAIVTMLKLL VAGEGRARTG VLIPIPQYPL YSAALAELDA VQVDYYLDEE RAWALDIAEL RRALCQARDR CCPRVLCVIN PGNPTGQVQT RECIEAVIRF AFEEGLFLMA DEVYQDNVYA EGSQFHSFKK VLTEMGPPYA TQQELASFHS VSKGYMGECG FRGGYVEVVN MDAEVQKQMA KLMSVRLCPP VPGQALMGMV VSPPTPSEPS FKQFQAERQE VLAELAAKAK LTEQVFNEAP GIRCNPVQGA MYSFPQIQLP LKAVQRAQDL GLAPDMFFCL CLLEETGICV VPGSGFGQQE GTYHFRMTIL PPMEKLRVLL EKLRHFHAKF THEYS | |
Greater than 90% as determined by SDS-PAGE. |
76282 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |