missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse 60S acidic ribosomal protein P0 Protein
A DNA sequence encoding the Mus musculus (Mouse) 60S acidic ribosomal protein P0, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6624-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_031501.1 | |
60S acidic ribosomal protein P0 Protein | |
Research Use Only | |
RPLP0 | |
Recombinant Protein | |
E. coli | |
MPREDRATWK SNYFLKIIQL LDDYPKCFIV GADNVGSKQM QQIRMSLRGK AVVLMGKNTM MRKAIRGHLE NNPALEKLLP HIRGNVGFVF TKEDLTEIRD MLLANKVPAA ARAGAIAPCE VTVPAQNTGL GPEKTSFFQA LGITTKISRG TIEILSDVQL IKTGDKVGAS EATLLNMLNI SPFSFGLIIQ QVFDNGSIYN PEVLDITEQA LHSRFLEGVR NVASVCLQIG YPTVASVPHS IINGYKRVLA LSVETEYTFP LTEKVKAFLA DPSAFAAAAP AAAATTAAPA AAAAPAKAEA KEESEESDED MGFGLFD | |
Greater than 80% as determined by SDS-PAGE. |
11837 | |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |