missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse 14-3-3 protein zeta / delta Protein
A DNA sequence encoding the Mus musculus (Mouse) 14-3-3 protein zeta/delta, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6899-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001240734.1 | |
14-3-3 protein zeta / delta Protein | |
Research Use Only | |
Ywhaz | |
Recombinant Protein | |
E. coli | |
MDKNELVQKA KLAEQAERYD DMAACMKSVT EQGAELSNEE RNLLSVAYKN VVGARRSSWR VVSSIEQKTE GAEKKQQMAR EYREKIETEL RDICNDVLSL LEKFLIPNAS QPESKVFYLK MKGDYYRYLA EVAAGDDKKG IVDQSQQAYQ EAFEISKKEM QPTHPIRLGL ALNFSVFYYE ILNSPEKACS LAKTAFDEAI AELDTLSEES YKDSTLIMQL LRDNLTLWTS DTQGDEAEAG EGGEN | |
Greater than 90% as determined by SDS-PAGE. |
22631 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |