missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Macaque MAPT / Tau Protein

A DNA sequence encoding the Macaca mulatta (Rhesus macaque) MAPT / Tau, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6344-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16022620

  • 860.00 EUR / 100µg
Estimated Shipment: 30-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

XP_014975440.1
MAPT / Tau Protein
100 μg
Not determined.
Full Length
Macaque
His-SUMO
Tris-based buffer, 50% glycerol
574327
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Mapt
Recombinant Protein
E. coli
AEPRQEFDVM EDHAGTYGLG DRKDQEGYTM LQDQEGDTDA GLKESPLQTP AEDGSEELGS ETSDAKSTPT AEDVTAPLVD ERAPGEQAAA QPHMEIPEGT TAEEAGIGDT PSLEDEAAGH VTQARMVSKS KDGTGSDDKK AKGADGKTKI ATPRGAAPPG QKGQANATRI PAKTPPAPKT PPSSATKQVQ RKPPPAEPTS ERGEPPKSGD RSGYSSPGSP GTPGSRSRTP SLPTPPAREP KKVAVVRTPP KSPSSAKSRL QTAPVPMPDL KNVKSKIGST ENLKHQPGGG KVQIINKKLD LSNVQSKCGS KDNIKHVPGG GSVQIVYKPV DLSKVTSKCG SLGNIHHKPG GGQVEVKSEK LDFKDRVQSK IGSLDNITHV PGGGNKKIET HKLTFRENAK AKTDHGAEIV YKSPVVSGDT SPRHLSNVSS TGSIDMVDSP QLATLADEVS ASLAKQGL
Videos
SDS
Documents
Certificates
Special Offers

Special Offers