missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Macaque IL10 / Interleukin-10 Protein
A DNA sequence encoding the Macaca mulatta (Rhesus macaque) IL10 / Interleukin-10, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9205-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001038192.1 | |
IL10 / Interleukin-10 Protein | |
Research Use Only | |
IL10 | |
Recombinant Protein | |
Yeast | |
SPGQGTQSEN SCTRFPGNLP HMLRDLRDAF SRVKTFFQMK DQLDNILLKE SLLEDFKGYL GCQALSEMIQ FYLEEVMPQA ENHDPDIKEH VNSLGENLKT LRLRLRRCHR FLPCENKSKA VEQVKNAFSK LQEKGVYKAM SEFDIFINYI EAYMTMKIQN | |
Greater than 90% as determined by SDS-PAGE. |
694931 | |
200 μg | |
Not determined. | |
Full Length | |
Macaque | |
His | |
Tris-based buffer, 50% glycerol |