missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Macaque IL-1 alpha / IL1A / IL1F1 Protein
A DNA sequence encoding the Macaca mulatta (Rhesus macaque) IL-1 alpha / IL1A / IL1F1 Protein, was expressed in the hosts and tags indicated.
309.00 EUR - 2985.00 EUR
Specifications
Accession Number | NP_001036222.1 |
---|---|
Gene ID (Entrez) | 700193 |
Name | IL-1 alpha / IL1A / IL1F1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16031620
|
enQuireBio™
QP6213-EC-100UG |
100 μg |
865.00 EUR
100µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16051620
|
enQuireBio™
QP6213-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16041620
|
enQuireBio™
QP6213-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061620
|
enQuireBio™
QP6213-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16071620
|
enQuireBio™
QP6213-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081620
|
enQuireBio™
QP6213-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001036222.1 | |
IL-1 alpha / IL1A / IL1F1 Protein | |
Not determined. | |
Full Length | |
Macaque | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
700193 | |
Research Use Only | |
IL1A | |
Recombinant Protein | |
E. coli | |
SAPFSFLSNM TYHFIRIIKH EFILNDTLNQ TIIRANDQHL TAAAIHNLDE AVKFDMGAYT SSKDDTKVPV ILRISKTQLY VSAQDEDQPV LLKEMPEINK TITGSETNFL FFWETHGTKN YFISVAHPNL FIATKHDNWV CLAKGLPSIT DFQILENQA | |
Greater than 90% as determined by SDS-PAGE. |