missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant M. tuberculosis Immunogenic protein MPT63 Protein

A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) Immunogenic protein MPT63, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8820-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16006661

  • 975.00 EUR / 100µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

WP_003409684.1
100 μg
Not determined.
Full Length
M. tuberculosis
His
Tris-based buffer, 50% glycerol
M. tuberculosis Immunogenic protein MPT63 Protein
Research Use Only
mpt63
Recombinant Protein
E. coli
AYPITGKLGS ELTMTDTVGQ VVLGWKVSDL KSSTAVIPGY PVAGQVWEAT ATVNAIRGSV TPAVSQFNAR TADGINYRVL WQAAGPDTIS GATIPQGEQS TGKIYFDVTG PSPTIVAMNN GMEDLLIWEP
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers