missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant M. tuberculosis ESAT-6-like protein EsxB Protein
A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) ESAT-6-like protein EsxB, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8759-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
WP_003399940.1 | |
1 mg | |
Not determined. | |
Full Length | |
M. tuberculosis | |
His | |
Tris-based buffer, 50% glycerol |
M. tuberculosis ESAT-6-like protein EsxB Protein | |
Research Use Only | |
esxB | |
Recombinant Protein | |
Yeast | |
AEMKTDAATL AQEAGNFERI SGDLKTQIDQ VESTAGSLQG QWRGAAGTAA QAAVVRFQEA ANKQKQELDE ISTNIRQAGV QYSRADEEQQ QALSSQMGF | |
Greater than 90% as determined by SDS-PAGE. |