missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant M. tuberculosis Ag85B Protein
A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) Ag85B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7004-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
WP_003409456.1 | |
50 μg | |
Not determined. | |
Full Length | |
M. tuberculosis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
M. tuberculosis Ag85B Protein | |
Research Use Only | |
fbpB | |
Recombinant Protein | |
E. coli | |
FSRPGLPVEY LQVPSPSMGR DIKVQFQSGG NNSPAVYLLD GLRAQDDYNG WDINTPAFEW YYQSGLSIVM PVGGQSSFYS DWYSPACGKA GCQTYKWETF LTSELPQWLS ANRAVKPTGS AAIGLSMAGS SAMILAAYHP QQFIYAGSLS ALLDPSQGMG PSLIGLAMGD AGGYKAADMW GPSSDPAWER NDPTQQIPKL VANNTRLWVY CGNGTPNELG GANIPAEFLE NFVRSSNLKF QDAYNAAGGH NAVFNFPPNG THSWEYWGAQ LNAMKGDLQS SLGAG | |
Greater than 90% as determined by SDS-PAGE. |