missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant M. tuberculosis Ag85A Protein

A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) Ag85A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8929-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16067201

  • 1405.00 EUR / 200µg
Estimated Shipment: 08-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

WP_003900759.1
200 μg
Not determined.
Partial (see sequence information for more details).
M. tuberculosis
His
Tris-based buffer, 50% glycerol
M. tuberculosis Ag85A Protein
Research Use Only
fbpA
Recombinant Protein
E. coli
YLQVPSPSMG RDIKVQFQSG GANSPALYLL DGLRAQDDFS GWDINTPAFE WYDQSGLSVV MPVGGQSSFY SDWYQPACGK AGCQTYKWET FLTSELPGWL QANRHVKPTG SAVVGLSMAA SSALTLAIYH PQQFVYAGAM SGLLDPSQAM GPTLIGLAMG DAGGYKASDM WGPKEDPAWQ RNDPLLNVGK LIANNTRVWV YCGNGKPSDL GGNNLPAKFL EGFVRTSNIK FQDAYNAGGG HNGVFDFPDS GTHSWEYWGA QLNAMKPDLQ RALGATPNT
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers