missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant M. tuberculosis Ag85A Protein
A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) Ag85A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8929-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
WP_003900759.1 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
M. tuberculosis | |
His | |
Tris-based buffer, 50% glycerol |
M. tuberculosis Ag85A Protein | |
Research Use Only | |
fbpA | |
Recombinant Protein | |
E. coli | |
YLQVPSPSMG RDIKVQFQSG GANSPALYLL DGLRAQDDFS GWDINTPAFE WYDQSGLSVV MPVGGQSSFY SDWYQPACGK AGCQTYKWET FLTSELPGWL QANRHVKPTG SAVVGLSMAA SSALTLAIYH PQQFVYAGAM SGLLDPSQAM GPTLIGLAMG DAGGYKASDM WGPKEDPAWQ RNDPLLNVGK LIANNTRVWV YCGNGKPSDL GGNNLPAKFL EGFVRTSNIK FQDAYNAGGG HNGVFDFPDS GTHSWEYWGA QLNAMKPDLQ RALGATPNT | |
Greater than 90% as determined by SDS-PAGE. |