missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza B (strain B / Singapore / 222 / 1979) NucleoProtein

A DNA sequence encoding the Influenza B virus (strain B/Singapore/222/1979) Influenza B (strain B/Singapore/222/1979) Nucleoprotein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7284-ye-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

Influenza B (strain B / Singapore / 222 / 1979) NucleoProtein
Research Use Only
NP
Recombinant Protein
Yeast
MSNMDIDGIN TGTIDKTPEE IISGTSGATR PIIRPATLAP PSNKRTRNPS PERATTSSEA DVGRKTQKKQ TPTEIKKSVY NMVVKLGEFY NQMMVKAGLN DDMERNLIQN AHAVERILLA ATDDKKTEFQ KKKNARDVKE GKEEIDHNKT GGTFYKMVRD DKTIYFSP
Greater than 90% as determined by SDS-PAGE.
200 μg
Not determined.
Extracellular Domain
Influenza
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers