missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza A H7N9 (Shanghai / 02 / 2013) Hemagglutinin Protein
A DNA sequence encoding the Influenza A virus (A/chicken/Jiangsu/1021/2013(H7N9)) Influenza A H7N9 (A/Shanghai/02/2013) Hemagglutinin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8948-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
YP_009118475.1 | |
Influenza A H7N9 (Shanghai / 02 / 2013) Hemagglutinin Protein | |
Research Use Only | |
HA | |
Recombinant Protein | |
E. coli | |
DKICLGHHAV SNGTKVNTLT ERGVEVVNAT ETVERTNIPR ICSKGKRTVD LGQCGLLGTI TGPPQCDQFL EFSADLIIER REGSDVCYPG KFVNEEALRQ ILRESGGIDK EAMGFTYSGI RTNGATSACR RSGSSFYAEM KWLLSNTDNA AFPQMTKSYK NTRKSPALIV WGIHHSVSTA EQTKLYGSGN KLVTVGSSNY QQSFVPSPGA RPQVNGLSGR IDFHWLMLNP NDTVTFSFNG AFIAPDRASF LRGKSMGIQS GVQVDANCEG DCYHSGGTII SNLPFQNIDS RAVGKCPRYV KQRSLLLATG MKNVPEIPKG RGLFGAIAGF IENGWEGLID GWYGFRHQNA QGEGTAADYK STQSAIDQIT GKLNRLIEKT NQQFELIDNE FNEVEKQIGN VINWTRDSIT EVWSYNAELL VAMENQHTID LADSEMDKLY ERVKRQLREN AEEDGTGCFE IFHKCDDDCM ASIRNNTYDH SKYREEAMQN RIQIDPVKLS SGYKDV | |
Greater than 90% as determined by SDS-PAGE. |
23104227 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Influenza | |
His-SUMO | |
Tris-based buffer, 50% glycerol |