missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza A H7N7 (Equine / Prague / 1 / 1956) NucleoProtein
A DNA sequence encoding the Influenza A virus (strain A/Equine/Prague/1/1956 H7N7) Influenza A H7N7 (strain A/Equine/Prague/1/1956) Nucleoprotein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7108-1mg
Additional Details : Weight : 0.01000kg
Specifications
Influenza A H7N7 (Equine / Prague / 1 / 1956) NucleoProtein | |
Research Use Only | |
NP | |
Recombinant Protein | |
E. coli | |
MASQGTKRPY EQMETGGERQ NATEIRASVG KMVGGIGRFY IQMCTELKLN DYEGRLIQNS ITIEKMVLSA FDERRNKYLE EHPNTGKDPK KTGGPIYRKR EGKWIRELIL YDKEEIRRIW RQANNGEDAT AGLTHLMIWH SNLNDATYQR TRALVRTGMD PRMCSLMQGS TLPRRSGAAG AAVKGIGTMV MELIRMIKRG INDRNFWRGE NGRKTRIAYE RMCNILKGKF QTAAQRAMMD QVRESRNPGN AEIEDLIFLA RSALILRGSV AHKSCLPACV YGLIVASGYD FEREGYSLVG VDPFKLLQNS QIFSLIRPNE NPAHKSQLVW MACHSAAFED LRVSSFIRGT KVIPRGQLST RGIQIASNEN METIDSNTLE LRSRYWAIRT KSGGNTSQQK ASAGQISVQP TFSVQRNLPF ERTTIMAAFT GNNEGRTSDM RTEIIRMMEN AKPDDVSFQG RGVFELSDEK ATNPIVPSFD MSKEGSYFFG DNAEEFDN | |
Greater than 80% as determined by SDS-PAGE. |
1 mg | |
Not determined. | |
Full Length | |
Influenza | |
His-SUMO | |
Tris-based buffer, 50% glycerol |