missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza A H3N2 (England / 878 / 1969) Hemagglutinin Protein
A DNA sequence encoding the Influenza A virus (strain A/England/878/1969 H3N2) Influenza A H3N2 (strain A/England/878/1969) Hemagglutinin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7283-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
Influenza A H3N2 (England / 878 / 1969) Hemagglutinin Protein | |
Research Use Only | |
HA | |
Recombinant Protein | |
E. coli | |
QDLPGNDNST ATLCLGHHAV PNGTLVKTIT NDQIEVTNAT ELVQSSSTGK ICNNPHRILD GINCTLIDAL LGDPHCDVFQ DETWDLFVER SKAFSNCYPY DVPDYASLRS LVASSGTLEF ITEGFTWTGV TQNGGSNACK RGPDSGFFSR LNWLTKSGST YPVLNVTMPN NDNFDKLYIW GVHHPSTNQE QTSLYVQASG RVTVSTRRSQ QTIIPNIGSR PWVRGLSSRI SIYWTIVKPG DVLVINSNGN LIAPRGYFKM RTGKSSIMRS DAPIDTCISE CITPNGSIPN DKPFQNVNKI TYGACPKYVK QNTLKLATGM RNVPEKQT | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Full Length | |
Influenza | |
His-SUMO | |
Tris-based buffer, 50% glycerol |