missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ZW10 interactor Protein
A DNA sequence encoding the Homo sapiens (Human) ZW10 interactor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7493-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001005413.1 | |
ZW10 interactor Protein | |
Research Use Only | |
ZWINT | |
Recombinant Protein | |
E. coli | |
MEAAETEAEA AALEVLAEVA GILEPVGLQE EAELPAKILV EFVVDSQKKD KLLCSQLQVA DFLQNILAQE DTAKGLDPLA SEDTSRQKAI AAKEQWKELK ATYREHVEAI KIGLTKALTQ MEEAQRKRTQ LREAFEQLQA KKQMAMEKRR AVQNQWQLQQ EKHLQHLAEV SAEVRERKTG TQQELDGVFQ KLGNLKQQAE QERDKLQRYQ TFLQLLYTLQ GKLLFPEAEA EAENLPDDKP QQPTRPQEQS TGDTMGRDPG VSFKAVGLQP AGDVNLP | |
Greater than 80% as determined by SDS-PAGE. |
11130 | |
10 μg | |
Not determined. | |
Full Length of BC020979 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |