missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ZRANB2 Protein

A DNA sequence encoding the Homo sapiens (Human) ZRANB2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6912-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16046710

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_005446.2
ZRANB2 Protein
Research Use Only
ZRANB2
Recombinant Protein
E. coli
MSTKNFRVSD GDWICPDKKC GNVNFARRTS CNRCGREKTT EAKMMKAGGT EIGKTLAEKS RGLFSANDWQ CKTCSNVNWA RRSECNMCNT PKYAKLEERT GYGGGFNERE NVEYIEREES DGEYDEFGRK KKKYRGKAVG PASILKEVED KESEGEEEDE DEDLSKYKLD EDEDEDDADL SKYNLDASEE EDSNKKKSNR RSRSKSRSSH SRSSSRSSSP SSSRSRSRSR SRSSSSSQSR SRSSSRERSR SRGSKSRSSS RSHRGSSSPR KRSYSSSSSS PERNRKRSRS RSSSSGDRKK RRTRSRSPES QVIGENTKQP
Greater than 90% as determined by SDS-PAGE.
9406
10 μg
Not determined.
Full Length of Isoform 2
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers