missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Zona pellucida sperm-binding protein 1
A DNA sequence encoding the Homo sapiens (Human) Zona pellucida sperm-binding protein 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6911-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001103824.1 | |
Zona pellucida sperm-binding protein 1 | |
Research Use Only | |
ZP3 | |
Recombinant Protein | |
E. coli | |
QPLWLLQGGA SHPETSVQPV LVECQEATLM VMVSKDLFGT GKLIRAADLT LGPEACEPLV SMDTEDVVRF EVGLHECGNS MQVTDDALVY STFLLHDPRP VGNLSIVRTN RAEIPIECRY PRQGNVSSQA ILPTWLPFRT TVFSEEKLTF SLRLMEENWN AEKRSPTFHL GDAAHLQAEI HTGSHVPLRL FVDHCVATPT PDQNASPYHT IVDFHGCLVD GLTDASSAFK VPRPGPDTLQ FTVDVFHFAN DSRNMIYITC HLKVTLAEQD PDELNKACSF SKPSNSWFPV EGSADICQCC NKGDCGTPSH SRRQPHVMSQ WSRSASRNRR HVTEEADVTV GPLIFLDRRG DHEVEQWALP SDTSV | |
Greater than 90% as determined by SDS-PAGE. |
7784 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |