missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Zinc finger protein 638 Protein

A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein 638, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6910-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16066700

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001014972.1
Zinc finger protein 638 Protein
Research Use Only
ZNF638
Recombinant Protein
E. coli
TFATLNTKGN EGDTVRDSIG FISSQVPEDP STLVTVDEIQ DDSSDLHLVT LDEVTEEDED SLADFNNLKE ELNFVTVDEV GEEEDGDNDL KVELAQSKND HPTDKKGNRK KRAVDTKKTK LESLSQVGPV NENVMEEDLK TMIERHLTAK TPTKRVRIGK TLPSEKAVVT EPAKGEEAFQ MSEVDEESGL KDSEPERKRK KTEDSSSGKS VASDVPEELD FLVPKAGFFC PICSLFYSGE KAMTNHCKST RHKQNTEKFM AKQRKEKEQN EAEERSSR
Greater than 90% as determined by SDS-PAGE.
27332
50 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers